EMAIL THIS PAGE TO A FRIEND
Chemosphere
Inhibition of animal acetyl-coenzyme A carboxylase by 2-(p-chlorophenoxy)-2-methylpropionic acid and 2-ethylhexanoic acid.
PMID 8938993
Abstract
Peroxisome proliferators are generally known as activators of fatty acid beta-oxidation which is one of degradation pathways. However, since it is unknown whether peroxisome proliferators have an effect on biosynthesis of fatty acid or not, we commenced to study if acetyl-CoA carboxylase, the rate-limiting enzyme of fatty acid biosynthesis, is inhibited by 2-(p-chlorophenoxy)-2-methylpropionic acid or 2-ethylhexanoic acid which is a typical peroxisome proliferator. Acetyl-CoA carboxylase(s) from rat, mouse, hamster, rabbit, dog and monkey were completely inhibited by 2-(p-chlorophenoxy)-2-methylpropionic acid or 2-ethylhexanoic acid at the concentration of 10(-3)M. For both compounds, there was not a large species difference in the inhibition. However, there was only several times difference between I50 values for the two compounds. The findings imply that inhibition of acetyl-CoA carboxylase by the chemicals leads to reduction of fatty acid biosynthesis when the chemicals are administered to the animals at high dose levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
197777
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
C10H11ClO3