EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Tyrothricin-containing oral tablets causing Clostridium Difficile-associated diarrhea.
PMID 8943986
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3000000 Tyrothricin, European Pharmacopoeia (EP) Reference Standard