EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Evaluation of fluorinated indole derivatives for electron-capture negative ionization mass spectrometry: application to the measurement of endogenous 5-methoxyindole-3-acetic acid in rat.
PMID 8946731
Abstract
A series of N-trifluoroacetyl/pentafluoropropionyl-O-trifluoroethyl/ pentafluoropropyl/heptafluorobutyl ester derivatives of 5-methoxyindole-3-acetic acid (5MIAA) were synthesized. Under electron-capture negative ionization conditions, the N-trifluoroacetyl derivatives were found to yield relatively abundant, analyte-specific M-. molecular ions and [M-HF]-., [M-HF-CF2CO]-. and [M-CF3CO]- fragment ions, while the N-pentafluoropropionyl derivatives yielded predominantly the reagent-specific pentafluoroacylium C2F5CO- ion. 5-[2H3]Methoxyindole-3-acetic acid was prepared in high yield by a new synthetic procedure and used as the internal standard in subsequent gas chromatographic/mass spectrometric analysis. Using the N-trifluoroacetyl-O-pentafluoropropyl ester derivative, femtomole to low picomole per gland/organ per g ml-1 levels of endogenous 5MIAA were identified and determined in the rat pineal gland, retina, whole brain and serum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M14935
5-Methoxy-3-indoleacetic acid, 98%
C11H11NO3