EMAIL THIS PAGE TO A FRIEND
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society
Contact allergy to polyvinylpyrrolidone/hexadecene copolymer previously reported.
PMID 8955492