EMAIL THIS PAGE TO A FRIEND
Drugs
Erdosteine.
PMID 8957158
Abstract
Erdosteine is a thiol derivative developed for the treatment of chronic obstructive bronchitis, including acute infective exacerbation of chronic bronchitis. Erdosteine contains 2 blocked sulfhydryl groups which are released following first-pass metabolism. The 3 active metabolites exhibit mucolytic and free radical scavenging activity. Erdosteine modulates mucus production and viscosity and increases mucociliary transport, thereby improving expectoration. It also exhibits inhibitory activity against the effects of free radicals produced by cigarette smoke. Clinical studies in patients with chronic obstructive lung disease have demonstrated the efficacy and tolerability of erdosteine. Erdosteine 300mg twice daily reduced cough (both frequency and severity) and sputum viscosity more quickly and more effectively than placebo and reduced the adhesivity of sputum more effectively than ambroxol 30mg twice daily. Co-administration of erdosteine and amoxicillin in patients with acute infective exacerbation of chronic bronchitis resulted in higher concentrations of the antibiotic in the sputum, leading to earlier and more pronounced amelioration of clinical symptoms compared with placebo. Erdosteine is associated with a low incidence of adverse events, most of which are gastrointestinal and generally mild.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1529
Erdosteine, ≥98% (HPLC)
C8H11NO4S2