EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Lethal and sublethal effects of azulene and longifolene to Microtox(R), Ceriodaphnia dubia, Daphnia magna, and Pimephales promelas.
PMID 8975804