EMAIL THIS PAGE TO A FRIEND
FEBS letters
BS-RNase tetramers: an example of domain-swapped oligomers.
PMID 8977132
Abstract
In the ribonuclease superfamily, dimericity is a unique feature of bovine seminal RNase (BS-RNase). In about two-thirds of native BS-RNase molecules, the two subunits interchange their N-terminal tails, thus generating domain-swapped dimers (MxM), which mostly responsible for enzyme biological activities and allostericity. Higher molecular weight BS-RNase oligomers can also be prepared [Libonati, M. (1969) Ital. J. Biochem. 18, 407-417.]. This paper reports on BS-RNase tetrameric derivatives which were isolated and enzymatically characterized. The data collected and the analysis of the crystal packing of MxM dimers suggested a structural model for tetramer assembly, in which the four subunits are enchained by multiple domain-swapping events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9630
Cytidine 2′:3′-cyclic monophosphate monosodium salt, ≥95% (HPLC)
C9H11N3NaO7P