EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
Endothelin-induced prostacyclin production in rat aortic rings is mediated by protein kinase C.
PMID 8981627
Abstract
Endothelin (ET) is a vasoconstrictor peptide released from endothelial cells that is known to cause prostaglandin release. The mechanism remains unclear. To determine whether the protein kinase C (PKC) signaling pathway is stimulated by endothelin, we pretreated rat aortic rings with either PKC activator or inhibitors and measured the release of prostacyclin (PGI2) by radioimmunoassay. ET (10(-9) M) produced a 10-fold increase in PGI2 release. Pretreatment with 10(-9) M of three different PKC inhibitors, 1-(5-isoquinolinesulfonyl)piperazine(CL), staurosporine, and 1-(5-isoquinolinesulfonyltmethyl)piperazine (H7), blocked ET-induced PGI2 release. ET-induced PGI2 release was also blocked by pretreatment with inhibitors of either phospholipase A2 7,7-dimethyleicosadienoic acid or trifluoromethyl ketone analogue) (10(-9) M) or cyclooxygenase (indomethacin) (10(-9) M). We conclude that ET activates PKC, which activates phospholipase A2, which liberates arachidonic acid, which increases PGI2 production and release.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8008
7,7-Dimethyl-(5Z,8Z)-eicosadienoic acid, ≥95%
C22H40O2