EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Modulation of the chaperone-like activity of bovine alpha-crystallin.
PMID 8986785
Abstract
The effects of pantethine, glutathione, and selected chemical reagents on the anti-aggregation activity of alpha-crystallin was evaluated. Protein aggregation was monitored by light scattering of solutions of denatured beta L-crystallin or alcohol dehydrogenase (ADH). The ratios of beta L-crystallin/alpha-crystallin and ADH/alpha-crystallin were adjusted so that partial inhibition of protein aggregation at 60 degrees C or 37 degrees C, respectively, was observed and modulation of the chaperone action of alpha-crystallin could be evaluated easily with selected endogenous metabolites. Enhancement of the anti-aggregation activity in the beta L-crystallin assay was strongest with pantethine, which appeared to interact with alpha-crystallin. Enhancement of the anti-aggregation activity in the ADH assay was strongest with glutathione which appeared to interact with ADH. The results indicated that the products of common metabolic pathways can modulate the chaperone-like effects of alpha-crystallin on protein aggregation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2125
D-Pantethine, analytical standard
C22H42N4O8S2