EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
In vitro susceptibility of clinical yeast isolates to fluconazole and terconazole.
PMID 8987951
Abstract
Fifty clinical yeast isolates, representing equally Candida albicans, Candida krusei, Candida parapsilosis, Candida tropicalis, and Torulopsis glabrata, were tested in vitro for their susceptibility to terconazole and fluconazole. The minimal inhibitory concentrations of terconazole and fluconazole were determined by use of a proposed standardized broth macrodilution assay. Also, the response of selected yeast isolates to 25 micrograms of either drug was measured by agarose disk diffusion experiments. For all species the minimum inhibitory concentrations for terconazole were significantly lower than those for fluconazole (p < 0.05). In fact, for each individual isolate the minimum inhibitory concentration of terconazole was consistently lower than that of fluconazole. Differences in the geometric mean of terconazole and fluconazole minimum inhibitory concentrations were largest among C. krusei and T. glabrata, followed by C. parapsilosis, C. tropicalis, and C. albicans, in order of decreasing difference. Disk diffusion experiments suggested that terconazole is a more effective fungistatic agent than fluconazole is. Terconazole may be more effective than fluconazole against yeast species other than C. albicans.