EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and protein kinase C inhibitory activities of balanol analogs with replacement of the perhydroazepine moiety.
PMID 9003521
Abstract
Balanol is a potent protein kinase C (PKC) inhibitor that is structurally composed of a benzophenone diacid, a 4-hydroxybenzamide, and a perhydroazepine ring. A number of balanol analogs in which the perhydroazepine moiety is replaced have been synthesized and their biological activities evaluated against both PKC and cAMP-dependent kinase (PKA). The results suggested that the activity and the isozyme/kinase selectivity of these compounds are largely related to the conformation about this nonaromatic structural element of the molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270253
4-Hydroxybenzamide, 98%
C7H7NO2