EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Response of three microbial test systems to pesticides.
PMID 9008061
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32014
Endrin, PESTANAL®, analytical standard
C12H8Cl6O
49032
Endrin, analytical standard
C12H8Cl6O