EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical applications
Sensitive analytical method for the novel H1-receptor antagonist HSR-609 in human plasma and urine by high-performance liquid chromatography with pre-column fluorescent derivatization.
PMID 9017466
Abstract
A simple and sensitive method for quantitation of HSR-609 (I) in human plasma and urine was developed using HPLC with the fluorescence labelling reagent 4-(N,N-dimethylaminosulfonyl)-7-N-piperazino-2,1,3-benzox adi azole (DBD-PZ). Compound I was extracted from human plasma and urine, and derivatized by reaction with DBD-PZ in the presence of Mukaiyama reagent A, an equimolar solution of 2,2'-dipyridyl disulfide (DPDS) and triphenylphosphine (TPP) in acetonitrile. The reaction mixture was cleaned up by liquid liquid extraction following the derivatization. The conjugate was analyzed by ion-pair-HPLC with fluorometric detection. The quantitation limits for I were 0.5 ng/ml in plasma and 5 ng/ml in urine. Using this method, plasma concentration and urinary excretion of I were studied after oral administration of I to human volunteers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93087
4-(N,N-Dimethylsulfamoyl)-7-piperazino-benzofurazan, for fluorescence, ≥99.0%
C12H17N5O3S