EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
The activities of four anticancer alkyllysophospholipids against Leishmania donovani, Trypanosoma cruzi and Trypanosoma brucei.
PMID 9023651
Abstract
The in-vitro activities of four anticancer alkyllysophospholipids, ET-18-OCH3 (edelfosine), hexadecylphosphocholine (miltefosine), ilmofosine and SRI 62-834, were determined with ED50 values for Leishmania donovani and Trypanosoma cruzi amastigotes between 0.2 and 5.0 microM and for Trypanosoma brucei trypomastigotes between 7.0 and 50.8 microM. In BALB/c mice miltefosine and ilmofosine were the most active compounds with ED50 values of 2.9 and 14.5 mg/kg x 5 doses against L. donovani and a suppressive effect on T. cruzi infections at 30 mg/kg x 5 doses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2409
Ilmofosine, ≥97% (NMR), semisolid (waxy)
C26H56NO5PS