EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Metabolism of amitriptyline with CYP2D6 expressed in a human cell line.
PMID 9041677
Abstract
1. Expressed human cytochrome P450 enzyme CPY2D6 was used to metabolize amitriptyline (AMI). It was established that CYP2D6 not only catalyzed ring 10-hydroxylation of AMI, but also mediated its N-demethylation to nortriptyline (NT), as well as the formation of 10-hydroxy-NT from NT. When the metabolism of AMI by CYP2D6 was repeated in the presence of quinidine, none of the metabolites, 10-hydroxy-AMI, NT and 10-hydroxy-NT, was formed. 2. Biochemical parameters of NT formation from AMI were determined, yielding Km = 47.48 +/- 1.32 microM; Vmax = 3.95 +/- 0.11 nmol/h/mg protein. The same parameters were calculated for the formation of 10-hydroxy-AMI (E + Z-isomers) from AMI, yielding Km = 10.70 +/- 0.20 microM; Vmax = 8.99 +/- 0.47 nmol/h/mg protein. 3. The formation of 10-hydroxy-NT from AMI proceeded primarily via NT and to a much lesser extent via 10-hydroxy-AMI. 4. Quantitative analyses of AMI and its metabolites were difficult to reproduce when the metabolites were analysed underivatized. Two derivatization procedures, acetylation and trifluoroacetylation, were employed to improve assay reproducibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A115
Amitriptyline metabolite, (±)-E-10-hydroxylated-, analytical standard
C20H23NO