EMAIL THIS PAGE TO A FRIEND
Phytochemistry
21'-Di-dehydro-deacetyllanatoside C, a biotransformation product of deacetyllanatoside C from senescent shoot cultures of Digitalis lanata.
PMID 9055448
Abstract
Feeding deacetyllanatoside C to senescent shoot cultures of Digitalis lanata resulted in the formation of a new product, which was isolated by semi-preparative HPLC. The molecular structure was elucidated by means of HPLC-mass spectrometry and NMR as 21'-di-dehydro-deacetyllanatoside C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19