EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact dermatitis due to emulsifiers.
PMID 9062756
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3386
Sorbitan sesquioleate, nonionic surfactant