EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Inhibitory effects of caffeic acid ethyl ester on H2O2-induced cytotoxicity and DNA single-strand breaks in Chinese hamster V79 cells.
PMID 9063980
Abstract
Cytotoxicity and DNA single-strand breaks caused by H2O2 were assessed by a colony formation assay and a DNA precipitation assay, respectively, with Chinese hamster V79 cells. In both assays, caffeic acid ethyl ester showed protective effects. The structure-activity relationship showed that the o-dihydroxy structure of caffeic acid ethyl ester was essential for the protective effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
320617
Ethyl 4-hydroxy-3-methoxycinnamate, 98%
C12H14O4