EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Use of the HPLC method for determination of glucopiranosides during ripening of faba bean seeds].
PMID 9064746
Abstract
Uridine (C9H12N2O6) was applied as an internal standard for the determination of glucopiranosides in faba bean seeds by the HPLC method. The relative UV response factors of vaccine or convicine to uridine were determined (1.12 and 0.79, respectively). The changes in the content of vaccine and convicine during ripening of faba bean seeds were followed. It was observed that with the increase in the seed dry matter content from 0.18 kg/kg to 0.78 kg/kg, the dry matter based content of vaccine decreased from 26.225 g/kg to 0.243 g/kg, and that of convicine from 9.051 g/kg to 0.094 g/kg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78260
Vicine, analytical standard
C10H16N4O7