EMAIL THIS PAGE TO A FRIEND
Journal of neurology, neurosurgery, and psychiatry
Unilateral auditory hallucinations in a boy with ipsilateral conductive hearing loss.
PMID 9069505
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1818
Molindone hydrochloride, ≥98% (HPLC), solid
C16H24N2O2 · HCl