EMAIL THIS PAGE TO A FRIEND
The Journal of toxicological sciences
Molecular cloning of mouse liver flavin containing monooxygenase (FMO1) cDNA and characterization of the expression product: metabolism of the neurotoxin, 1,2,3,4-tetrahydroisoquinoline (TIQ).
PMID 9076656
Abstract
A mouse liver cDNA clone, MFMO1, coding for a flavin-containing monooxygenase (FMO) was isolated. This cDNA clone encoded a protein of 532 amino acids. Based upon its predicted amino acid sequence, this clone was assumed to belong to the FMO1 subfamily. The deduced amino acid sequence showed 94, 84, 83, and 83% identity with FMO1s of rats, pigs, rabbits and humans, respectively, while it showed only 50-59% identity with human FMO3 and FMO4, rabbit FMO2, FMO3, FMO4 and FMO5, and guinea-pig FMO2. RNA blot analysis showed that the mouse FMO1 was also expressed in the lung and kidney and to lesser extents in the heart, spleen, testis and brain. Mouse FMO1 expressed in yeast showed activities of thiobenzamide S-oxidation, and NADPH oxidation associated with the S- or N-oxidation of chlorpromazine, N,N-dimethylaniline, N,N-dimethyl-hydrazine, imipramine, nicotine, thioacetamide, thiourea and trimethylamine. Moreover, 1,2,3,4-tetrahydroisoquinoline (TIQ), a substance known to induce a parkinsonism-like syndrome in monkeys, was also metabolized by the mouse FMO1. The K(m) values for chlorpromazine, imipramine and TIQ were determined to be 2,4, 16.0, 435 mM, respectively. This is the first report to show that an expressed FMO can metabolize a neurotoxin, TIQ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45328
ANTU, PESTANAL®, analytical standard
C11H10N2S