EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Evaluation of potential immunotoxicity of pesticides in human lymphocyte tests].
PMID 9081885
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36194
Phosalone, PESTANAL®, analytical standard
C12H15ClNO4PS2
PS682
Phosalone, analytical standard
C12H15ClNO4PS2