EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Identification of beta-L-gulose as the sugar moiety of the main polar lipid Thermoplasma acidophilum.
PMID 9084501
Abstract
The main polar lipid (MPL) of Thermoplasma acidophilum has been purified and its structure determined. NMR, mass spectrometry, and capillary gas chromatography-mass spectrometry experiments have shown that the previously unidentified sugar moiety of MPL is the rare sugar L-gulose. MPL is thus a tetraether lipid with cyclopentane rings and head groups of phosphoglycerol, as previously reported, and beta-L-gulopyranose. Further, MPL is also the dominant lipid found in lipid extracts from another species of the Thermoplasma genus, T. volcanium, suggesting that L-gulose may represent a dominant sugar moiety of the polar lipids biosynthesized by this archaeobacterial genus. Minor phospholipids were tentatively identified as diether and hydroxydiether analogs of phosphatidylglycerol, and phosphatidylinositol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56224
L-(+)-Gulose, ≥98.0% (HPLC)
C6H12O6