EMAIL THIS PAGE TO A FRIEND
Human mutation
G-->T transition at cDNA nt 110 (K37Q) in the PKLR (pyruvate kinase) gene is the molecular basis of a case of hereditary increase of red blood cell ATP.
PMID 9090535