EMAIL THIS PAGE TO A FRIEND
Steroids
Estrone sulfonates as inhibitors of estrone sulfatase.
PMID 9090794
Abstract
In our continuing quest to design efficient inhibitors of estrone sulfatase activity and to assess the recognition of estrone sulfate surrogates by estrone sulfatase, we synthesized and evaluated several sulfonate derivatives of 5,6,7,8-tetrahydronaphth-2-ol and estrone. 5,6,7,8-Tetrahydronaphth-2-methanesulfonate (11), and 5,6,7,8-tetrahydronaphth-2-(p-toluene)sulfonate (12) were found not to inhibit estrone sulfatase activity; estrone-3-methane-sulfonate (5), estrone-3-ethanesulfonate (6), estrone-3-butanesulfonate (7), and estrone-3-[(+)10-camphor]sulfonate (8) all weakly inhibited estrone sulfatase, and the best inhibitor, from this class of compounds, was estrone-3-(p-toluene)sulfonate (9). At 10 microM, it inhibited estrone sulfatase activity by 91%. These results emphasize some of the requirements needed for high-affinity binding to the enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180459
5,6,7,8-Tetrahydro-2-naphthol, 98%
C10H12O