EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Reductive cleavage of permethylated polysaccharides with borane-methyl sulfide complex and butyltin trichloride.
PMID 9090811
Abstract
Several per-O-methylated monosaccharides and polysaccharides were used as models in an attempt to identify more convenient reagents for accomplishing reductive cleavage of glycosidic linkages. Included in the model studies were methyl alpha- and beta-D-glucopyranoside, methyl alpha- and beta-D-mannopyranoside, and methyl alpha- and beta-D-ribofuranoside. These studies led to the identification of a new promoter, butyltin trichloride, for carrying out reductive cleavage when borane-methyl sulfide complex was used as the reducing agent. These reagents were found to accomplish the reductive cleavage of per-O-methylated amylose, cellulose, and pullulan to give only the expected derivatives of 1,5-anhydro-D-glucitol. These reagents also accomplished reductive cleavage of per-O-methylated insulin to give only the expected derivatives of 2,5-anhydro-D-mannitol and 2,5-anhydro-D-glucitol. Reductive cleavage using these reagents is easy to perform, and subsequent acetylation of the products is readily accomplished in situ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201057
Butyltin trichloride, 95%
C4H9Cl3Sn