EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Mutagenic carcinogens and noncarcinogens in transgenic mice.
PMID 9105788
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148113
2,6-Diaminotoluene, 97%
C7H10N2
45921
2-Methyl-m-phenylenediamine, analytical standard
C7H10N2