EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Anthelmintic resistance of trichostrongylids in sheep in the highveld of Zimbabwe.
PMID 9106960
Abstract
A survey was conducted on the occurrence of anthelmintic resistance of trichostrongylids on commercial sheep farms in the highveld of Zimbabwe. On ten farms the efficacy of fenbendazole, levamisole and rafoxanide was tested by a faecal egg count reduction test. Benzimidazole resistance was additionally examined using an egg hatch assay with thiabendazole. Results of the faecal egg count reduction tests and larval differentiations showed fenbendazole resistance of Haemonchus sp. on all investigated farms. Resistance of Haemonchus sp. against rafoxanide was demonstrated on all farms with reliable egg counts. Levamisole resistance of Haemonchus sp. was found on most farms but 2 farms showed an efficacy of 100%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34042
Rafoxanide, PESTANAL®, analytical standard
C19H11Cl2I2NO3