EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Evolution of bacterial genomes.
PMID 9111921
Abstract
This review examines evolution of bacterial genomes with an emphasis on RNA based life, the transition to functional DNA and small evolving genomes (possible plasmids) that led to larger, functional bacterial genomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D26202
3,5-Diamino-1,2,4-triazole, 98%
C2H5N5