EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Reproductive toxicology. Di-n-propylphthalate.
PMID 9114313
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290602
Dipropyl phthalate, 98%
C14H18O4
45624
Dipropyl phthalate, PESTANAL®, analytical standard
C14H18O4