EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Reproductive toxicology. 2-Chloronitrobenzene.
PMID 9114328
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45926
1-Chloro-2-nitrobenzene, analytical standard
C6H4ClNO2
185760
1-Chloro-2-nitrobenzene, 99%
C6H4ClNO2