EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Reproductive toxicology. 1-Chloro-2-propanol.
PMID 9114330
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292087
1-Chloro-2-propanol, 70%
C3H7ClO