EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Reproductive toxicology. 4-Vinylcyclohexene.
PMID 9114365
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94950
4-Vinyl-1-cyclohexene, analytical standard
C8H12