EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Northern bobwhite egg hatchability and chick immunocompetence following a field application of clopyralid.
PMID 9115146
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36758
Clopyralid, PESTANAL®, analytical standard
C6H3Cl2NO2