EMAIL THIS PAGE TO A FRIEND
Rivista di biologia
Two-step hydrolysis of amygdalin in molds.
PMID 9122587
Abstract
Mucor circinelloides LU M40 and Penicillium aurantiogriseum P 35, characterized by extracellular beta-glucosidase activity on cyanogenic glycosides, hydrolyse amygdalin by a two-step reaction mechanism being the first step of hydrolysis, from amygdalin to prunasin, very rapid (15 min) and the second one, from prunasin to mandelonitrile, much slower (120 min).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00173
Prunasin, ≥90% (LC/MS-ELSD)
C14H17NO6