EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Apoptotic cell death in activated monocytes following incorporation of clodronate-liposomes.
PMID 9129215
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3440 Polyoxyethylene (40) stearate