EMAIL THIS PAGE TO A FRIEND
Psychiatria polska
[Sulpiride: an atypical neuroleptic with a wide spectrum of therapeutic claims].
PMID 9132763
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8010
(±)-Sulpiride
C15H23N3O4S
S2190000
Sulpiride, European Pharmacopoeia (EP) Reference Standard
C15H23N3O4S