EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Studies on cardiac ingredients of plants. XIII: Chemical modification of gitoxin to cardiotonic compounds without vascular effect.
PMID 9145499
Abstract
Nitrated gitoxins (4) and bufotoxin homologues with various lengths of alkyl chain at C-3 of the steroid nucleus (10) were prepared from gitoxin (1). The pharmacological activities of the resulting compounds (4 and 10) were evaluated by measurement of inhibitory effect on NA+, K(+)-adenosine triphosphatase (ATPase) prepared from dog kidney, positive inotropic effect (PIE) on isolated guinea-pig papillary muscle preparations, and the effect on smooth muscle using the mesenteric artery from spontaneously hypertensive rats. Most of the compounds showed a smaller contractile effect on the arterial muscle. Among these compounds, gitoxin 3"-nitrate (4g) exhibited the most desirable biological activities, such as PIE comparable to that of 1, 1.25 times wider concentration-dependent range than 1, and lack of contractile activity on vascular muscle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64107
Gitoxin, ≥99% (HPLC)
C41H64O14