EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
Effects of noradrenaline on frequency tuning of rat auditory cortex neurons.
PMID 9153591
Abstract
The selectivity of rat auditory cortex neurons for pure tone frequency was studied during and after ionophoretic application (5-40 nA) of noradrenaline in urethane-anaesthetized rats. The dominant effect induced by noradrenaline was a significant decrease in spontaneous (93/268 cells) and evoked activity (133/268 cells) which outlasted the application. In the whole population of cells (n = 268) the signal-to-noise ratio, computed using as the signal either the mean evoked response or the response at the best frequency, was unchanged during noradrenaline application. It was significantly increased only for cells showing significantly decreased spontaneous activity, and was significantly decreased for cells showing increased spontaneous activity. Frequency selectivity was significantly increased for the whole population during and after noradrenaline application. It was also significantly increased for cells showing significantly decreased evoked activity, and was significantly decreased for cells showing increased evoked activity. The noradrenaline-induced inhibition was not blocked by propranolol (beta antagonist); it was blocked by prazosin (alpha1 antagonist) and partly mimicked by phenylephrine (alpha1 agonist). GABA, which also inhibited spontaneous and evoked activity, slightly increased the signal-to-noise ratio and significant increased frequency selectivity. However, when noradrenaline was ejected in the presence of bicuculline at doses that were able to block GABAergic inhibition, the inhibitory effects of noradrenaline on spontaneous and evoked activity were still observed. The possible function of noradrenaline-induced inhibitions in sensory cortices is briefly discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2760
(−)-Isoproterenol (+)-bitartrate salt, powder
C11H17NO3 · C4H6O6