EMAIL THIS PAGE TO A FRIEND
Canadian journal of psychiatry. Revue canadienne de psychiatrie
Re: Rapid stabilization at home.
PMID 9161769
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F100
Fluspirilene
C29H31F2N3O