EMAIL THIS PAGE TO A FRIEND
International journal of legal medicine
Acute fatal poisoning with dichlorophen.
PMID 9168328
Abstract
A case is presented involving an acute fatality resulting from self-administered dichlorophen, a chlorophenol fungicide. The compound was quantified using gas chromatography/mass spectrometry after extraction with methyl-tert-butyl ether, derivatization by methylation and separation on a HP5-MS capillary column. The blood concentration was 9.77 mg/l and other drugs, including ethanol, were not detected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
133221
Bis(5-chloro-2-hydroxyphenyl)methane, 95%
C13H10Cl2O2
35992
Dichlorophene, PESTANAL®, analytical standard
C13H10Cl2O2