EMAIL THIS PAGE TO A FRIEND
Leukemia & Lymphoma
Molecular diagnosis and clinical relevance of t(9;22), t(4;11) and t(1 ;19) chromosome abnormalities in a consecutive group of 141 adult patients with acute lymphoblastic leukemia
PMID 9172811