EMAIL THIS PAGE TO A FRIEND
Nature Medicine
Restoration of the growth suppression function of mutant p53 by a synthetic peptide derived from the p53 C-terminal domain.
PMID 9176489
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25-006
p53 Activating Ligand