EMAIL THIS PAGE TO A FRIEND
Journal of Oral Pathology & Medicine
Histochemical and immunohistochemical localisation of elastic system fibres in focal reactive overgrowths of oral mucosa.
PMID 9176788
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB424
Anti-PCNA Antibody, clone PC10