EMAIL THIS PAGE TO A FRIEND
Steroids
Allylic nitration of 3 beta-sitosterol and cholesterol acetate: preparation of 7-nitro derivatives.
PMID 9185299
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151114
Cholesteryl acetate, 97%
C29H48O2