EMAIL THIS PAGE TO A FRIEND
FEBS letters
Postimport methylation of the small subunit of ribulose-1,5-bisphosphate carboxylase in chloroplasts.
PMID 9188792
Abstract
Electron impact mass spectronomy analysis of the amino-terminal amino acid of the small subunit (SSU) of ribulose-1,5-bisphosphate carboxylase (Rubisco) showed that the amino-terminal methionine residue is post-translationally modified to N-methyl-methionine. Modification of the amino-terminal methionine residue was found in mature SSU proteins from the dicotyledonous plants pea and spinach as well as the monocotyledonous plants barley and corn. SSU methyltransferase is a soluble protein in the chloroplast stroma and accepts heterologously expressed non-methylated SSU as a substrate using S-adenosylmethionine as methyl-group donor. We show that this modification occurs after post-translational uptake of the precursor form of SSU into chloroplasts and processing to its mature size. This reaction represents a new step in the import and assembly pathway of Rubisco holoenzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T28355
N,N′-Diphenylthiourea, 98%
C13H12N2S