EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[The characteristics of extracting 4-methyl-2,6-dinitrophenol and 2-(1-methylpropyl)-4,6-dinitrophenol from aqueous solutions].
PMID 9198983
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45451
Dinobuton, PESTANAL®, analytical standard
C14H18N2O7