EMAIL THIS PAGE TO A FRIEND
Cell
Cocrystal structure of the messenger RNA 5' cap-binding protein (eIF4E) bound to 7-methyl-GDP.
PMID 9200613
Abstract
The X-ray structure of the eukaryotic translation initiation factor 4E (eIF4E), bound to 7-methyl-GDP, has been determined at 2.2 A resolution. eIF4E recognizes 5' 7-methyl-G(5')ppp(5')N mRNA caps during the rate-limiting initiation step of translation. The protein resembles a cupped hand and consists of a curved, 8-stranded antiparallel beta sheet, backed by three long alpha helices. 7-methyl-GDP binds in a narrow cap-binding slot on the molecule's concave surface, where 7-methyl-guanine recognition is mediated by base sandwiching between two conserved tryptophans, plus formation of three hydrogen bonds and a van der Waals contact between its N7-methyl group and a third conserved tryptophan. The convex dorsal surface of the molecule displays a phylogenetically conserved hydrophobic/acidic portion, which may interact with other translation initiation factors and regulatory proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5883
7-Methylguanosine 5′-diphosphate sodium salt, ≥92.5%
C11H17N5O11P2 · xNa+