EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Hepatic hyperplasia and cancer in rats: metabolic alterations associated with cell growth.
PMID 9207284
Abstract
We showed previously that the peroxisome proliferators di(2-ethylhexyl)phthalate (DEHP), clofibrate, and 4-chloro-6-(2,3 xylidino)-2-pyrimidinylthio (N-beta-hydroxyl)acetamide (BR931) alter hepatic sex steroid metabolism and receptor expression during induction of hepatic hyperplasia and hepatocellular carcinoma (HCC) in rats. The aim of this study was to identify metabolic changes associated with cell growth during hyperplasia and HCC. Hepatic hyperplasia was induced in male rats by a diet containing DEHP and clofibrate for 3-60 days. HCC was induced by feeding a diet containing BR931, a more potent hepatocarcinogen, for 10 months. Cholesterol biosynthesis was depressed in hyperplastic livers but increased in HCC. Glucose-6-phosphate dehydrogenase (G6PD) activity was inhibited in hyperplastic liver as well as in HCC, whereas malic enzyme activity increased severalfold. Protein and messenger RNA (mRNA) levels for both G6PD and malic enzyme increased in hyperplastic livers and HCC. mRNA levels for 3-hydroxy-3-methylglutaryl-coenzyme A reductase decreased in hyperplasia and increased in HCC, whereas low-density lipoprotein receptor mRNA increased in hyperplasia and decreased in HCC. Neoplastic cells acquire a growth advantage by their capacity to synthesize cholesterol and obtain reduced nicotinamide adenine dinucleotide phosphate by the malic enzyme pathway when G6PD activity is inhibited by peroxisome proliferators.