EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical sciences and applications
Determination of cocaine and its metabolites in serum microsamples by high-performance liquid chromatography and its application to pharmacokinetics in rats.
PMID 9210433
Abstract
A single-solvent extraction step high-performance liquid chromatographic method is described for quantitating cocaine and its three metabolites in rat serum microsamples (50 microl). The separation used a 2.1-mm I.D. reversed-phase Brownlee C18 column with an isocratic mobile phase consisting of methanol-acetonitrile-25.8 mM sodium acetate buffer, pH 2.2, containing 1.29-10(-4) M tetrabutylammonium phosphate (12.5:10:77.5, v/v/v). The detection limit was 2.5 ng/ml for all the compounds using an ultraviolet detector operated at 235 nm. The method was used to study the pharmacokinetics of cocaine after an intravenous (i.v.) bolus dose (4 mg/kg).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B142
Benzoylnorecgonine, analytical standard
C15H17NO4